Learn More
Cayman Chemical (S)-MG132, 133407-82-6, 1 mg
SDP
Supplier:  Cayman Chemical 100126281
Encompass_Preferred
A potent, reversible, and cell permeable proteasome inhibitor; inhibits cell growth in B16 and IPC227F cells with IC50 values of 42 and 77 nM, respectively; inhibits NF-κB activation, sensitizing a variety of carcinoma cell lines to apoptosis
Catalog No. NC9038428