Learn More
Selleck Chemical LLC MG149
SDP
Supplier:  Selleck Chemical LLC S747610MM/1ML
Encompass_Preferred
MG149 (Tip60 HAT inhibitor) is a potent histone acetyltransferase inhibitor with IC50 of 74 M and 47 M for Tip60 and MOF respectively
Catalog No. 50-000-03559