Learn More
Selleck Chemical LLC MG149 5mg
SDP
Supplier:  Selleck Chemical LLC S74765MG
Encompass_Preferred
MG149 (Tip60 HAT inhibitor) is a potent histone acetyltransferase inhibitor with IC50 of 74 ?M and 47 ?M for Tip60 and MOF,respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3277