Learn More
Medchemexpress LLC MGD-28 | 2991818-13-2 | 99.7% | 595.67 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY168615100MG
Encompass_Preferred
MGD-28 is a potent, orally active, Cereblon (CRBN)-dependent IKAROS protein degrader that degrades IKZF1 (DC50=3.8 nM), IKZF2 (DC50=56.3 nM), and IKZF3 (DC50=7.1 nM) in a dose-dependent manner. In addition, MGD-4 also degrades CK1α (DC50=7.8 nM). MGD-28 has antiproliferative activity and can be used in multiple myeloma research.
  • Potent
  • Orally active
  • Cereblon (CRBN)-dependent IKAROS protein degrader
  • Degrades IKZF1, IKZF2, and IKZF3 in a dose-dependent manner
  • Degrades CK1α
  • Antiproliferative activity
  • Can be used in multiple myeloma research
Catalog No. 50-003-90404