Learn More
Selleck Chemical LLC MGH-CP1-5mg
SDP
Supplier:  Selleck Chemical LLC S97355MG
Encompass_Preferred
MGH-CP1 is a potent and selective inhibitor of transcriptional enhanced associate domain (TEAD) palmitoylation. MGH-CP1 exhibits dose-dependent and potent inhibition of TEAD2/4 auto-palmitoylation in vitro with IC50 of 710 nM and 672 nM, respectively.
Catalog No. 50-238-9928