Learn More
Enzo Life Sciences MHC Class I monoclonal antibody (W6/32) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805711C100
Encompass_Preferred
Clone: W6/32. Isotype: IgG2a. Immunogen: Membrane of human tonsil cells. Species reactivity: Human, Monkey, Cat, Bovine. Specificity: Recognizes virtually all nucleated human cells. Suitable as positive control for HLA tissue typing and crossmatching. Applications: ELISA, Flow Cytometry, ICC, IHC (FS), IP, WB, FUNC. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.4. Sterile filtered. Contains no preservatives.
Catalog No. 50-201-0065