Learn More
Apexbio Technology LLC MHC class II antigen (45-57) [Homo sapiens] 25mg
SDP
Supplier:  Apexbio Technology LLC A108125
Encompass_Preferred
MHC class II antigen 45-57 is a synthetic peptide derived from human major histocompatibility complex class II molecules It is designed to facilitate studies on antigen presentation by MHC class II molecules which bind extracellularly derived peptide antigens and present them to CD4 helper T lymphocytes thereby initiating adaptive immune responses MHC class II antigen 45-57 exerts its biological activity primarily by serving as a model antigenic peptide for investigating MHC II-mediated antigen presentation and T-cell activation Based on these properties MHC class II antigen 45-57 holds research potential in studies of MHC II-mediated immune responses T-cell activation and the cellular immune mechanisms involved in pathogen defense and autoimmune diseases
Catalog No. 50-000-93662