Learn More
Medchemexpress LLC MHY1485 | 326914-06-1 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0795200MG
Encompass_Preferred
MHY1485 is a potent, cell-permeable mTOR activator that targets the ATP domain of mTOR. It inhibits autophagy by suppressing the fusion between autophagosomes and lysosomes.
  • Inhibits GCDC-induced autophagic activity in HCC cells
  • Increases LC3II/LC3I ratio in rat liver Ac2F cells
  • Dose-dependently increases phosphorylation of mTOR at Ser2448 and upregulates 4E-BP1 phosphorylation in Ac2F cells
  • Blocks autophagy signaling induced by follicle-stimulating hormone (FSH) in female ICR mice
  • Increases p-mTOR and p-S6K1 expression levels in mice
Catalog No. 50-004-09494