Learn More
TARGETMOL CHEMICALS INC MI-136 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T688910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MI-136 inhibits expression of androgen receptor (AR) target genes that DHT induced. Purity 98.63%
Catalog No. 50-288-0471