Learn More
Medchemexpress LLC MI-1851 | 2417283-44-2 | C34H53N15O6 | 100 UG
SDP
Supplier:  Medchemexpress LLC HY150737100UG
Encompass_Preferred
MI-1851 is a potent furin inhibitor that prevents the proteolytic processing of the S protein of SARS-CoV-2 by endogenous flavoprotease in HEK293 cells and exhibits antiviral activity. It also inhibits the spread and proliferation of SARS-CoV-2 in Calu-3 cells and replication of DENV-2 and WNV in huh-7 cells.
  • Prevents proteolytic processing of S protein of SARS-CoV-2.
  • Exhibits antiviral activity.
  • Strongly inhibits the spread of SARS-CoV-2 in Calu-3 cells.
  • Inhibits DENV-2 and WNV replication.
  • Does not affect PHH cell viability.
Catalog No. 50-003-74044