Learn More
Abcam MI 2, MALT1 inhibitor, 50MG
Supplier:  Abcam AB14504750MG
Encompass_Preferred
MW 455.73 g/mol, Purity >98%. Cell-permeable, irreversible MALT1 inhibitor (IC₅₀ = 5.8 μM). Suppresses protease function, decreases NF-κB activity and inhibits cell proliferation. Antitumor agent. Active in vitro, in vivo and ex vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5450