Learn More
Medchemexpress LLC HY-136360 5mg Medchemexpress, MI-3454 CAS:2134169-43-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1363605MG
Encompass_Preferred
Medchemexpress, HY-136360 5mg MI-3454 CAS:2134169-43-8 MI-3454 is an orally active, highly potent and selective menin-MLL1 interaction inhibitor with an IC50 of 0.51 nM. MI-3454 inhibits proliferation, induces differentiation and complete remission or regression of leukemia in mouse models of MLL1-rearranged or NPM1-mutated leukemia through downregulation of key genes involved in leukemogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0425