Learn More
Medchemexpress LLC Mi-463 | 1628317-18-9 | 99.7% | 484.54 | C24H23F3N6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1980910MG
Encompass_Preferred
MI-463 is a potent, orally bioavailable small-molecule inhibitor of the menin-MLL protein-protein interaction, provided for research use. It is supplied as a solid with analytical data and formulation guidance to support in vitro and in vivo studies.
  • Potent, orally bioavailable inhibitor of the menin-MLL interaction.
  • High reported purity for reliable experimental results.
  • Well-characterized molecular weight for dosing calculations.
  • Soluble in DMSO with established in vivo formulation protocols.
  • Long-term storage conditions specified for powder and solutions.
  • Multiple pack sizes available to suit study scale.
Catalog No. 50-002-97543