Learn More
Selleck Chemical LLC MI-463
SDP
Supplier:  Selleck Chemical LLC S781625MG
Encompass_Preferred
MI-463 is a potent inhibitor of Menin-MLL interaction with an IC50 value of 15 3 nM
Catalog No. 50-000-01057