Learn More
Selleck Chemical LLC MI-503 2mg 1857417-13-0
SDP
Supplier:  Selleck Chemical LLC S78172MG
Encompass_Preferred
MI-503 is a potent and selective Menin-MLL inhibitor with IC50 of 14.7 nM. It shows pronounced growth suppressive activity in a panel of human MLL leukemia cell lines(GI50 at 250 nM-570 nM range), but only a minimal effect in human leukemia cell lines without MLL translocations. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5299