Learn More
Medchemexpress LLC HY-16925 2mg Medchemexpress, MI-503 CAS:1857417-13-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY169252MG
Encompass_Preferred
Medchemexpress, HY-16925 2mg MI-503 CAS:1857417-13-0 MI-503 is a highly potent and orally bioavailable small molecule inhibitor of the menin-mLL interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9219