Learn More
Medchemexpress LLC MI-773 | 1303607-07-9 | 96.3% | 562.50 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1749350MG
Encompass_Preferred
MI-773 is a potent MDM2-p53 protein-protein interaction (PPI) inhibitor with high binding affinity against MDM2 (Kd=8.2 nM) and has antitumor activity.
  • Potent MDM2-p53 protein-protein interaction (PPI) inhibitor
  • High binding affinity against MDM2 (Kd=8.2 nM)
  • Antitumor activity
  • Available in solid and solution forms
  • Soluble in DMSO (≥ 53 mg/mL)
Catalog No. 50-003-86790