Learn More
Medchemexpress LLC MI-773 | 1303607-07-9 | MFCD28168022 | 96.3% | 562.50 | C29H34Cl2FN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1749310MG
Encompass_Preferred
MI-773 is a small-molecule inhibitor of the MDM2-p53 protein-protein interaction with high binding affinity for MDM2 and reported antitumor activity in preclinical studies. It is provided as a research-grade compound for use in cancer biology and p53 pathway experiments.
  • High-affinity MDM2 binding (reported Kd 8.2 nM).
  • Reported antitumor activity in preclinical studies.
  • Provided as a research-grade compound suitable for in vitro and in vivo studies.
  • Available in small mg-scale quantities for research use.
  • Molecular weight 562.50 and formula C29H34Cl2FN3O3 for reference.
Catalog No. 50-002-98531