Learn More
Medchemexpress LLC Mianserin | 24219-97-4 | 99.4% | 50 MG
Supplier: Medchemexpress LLC HYB018850MG
Mianserin is an orally active H1 receptor antagonist that can activate κ-opioid and octopamine receptors. It increases ERK1/2 and CREB phosphorylation, and antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation. It modulates social and exploratory behavior and raises electroconvulsive thresholds. Mianserin is used for research of neurological diseases such as depression and epilepsy.
- Orally active H1 receptor antagonist
- Activates κ-opioid receptor
- Activates octopamine receptor
- Increases ERK1/2 and CREB phosphorylation
- Antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation
- Modulates social and exploratory behavior
- Raises electroconvulsive thresholds
- Useful for research of neurological diseases like depression and epilepsy