Learn More
Medchemexpress LLC Mianserin | 24219-97-4 | 99.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB01885MG
Encompass_Preferred
Mianserin (Mianserine) is an orally active H1 receptor antagonist. It can activate κ-opioid receptors and octopamine receptors. Mianserin increases ERK1/2 and CREB phosphorylation and antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation. It modulates social and exploratory behavior and raises electroconvulsive thresholds. It is used in research for neurological diseases such as depression and epilepsy.
  • Orally active H1 receptor antagonist
  • Activates κ-opioid and octopamine receptors
  • Increases ERK1/2 and CREB phosphorylation
  • Modulates social and exploratory behavior
  • Raises electroconvulsive thresholds
  • Used in research for neurological diseases such as depression and epilepsy
Catalog No. 50-004-05769