Learn More
Apexbio Technology LLC Mianserin HCl 21535-47-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A17965.1
Encompass_Preferred
Mianserin HCl (CAS 21535-47-7) is a small molecule that functions as a non-selective antagonist at 5-hydroxytryptamine (5-HT) 2 receptor subtypes and exhibits moderate affinity for 5-HT6 receptors Through its antagonistic activity at these serotonergic receptors Mianserin HCl modulates neurotransmission pathways implicated in mood regulation The compound is commonly utilized in neuropharmacological studies to investigate serotonergic signaling and has established utility as an antidepressant agent in clinical research
Catalog No. 50-000-95652