Learn More
Medchemexpress LLC Mianserin hydrochloride | 21535-47-7 | 99.95% | 1 ML
Supplier: Medchemexpress LLC HYB0188A10MM/1ML
Mianserin hydrochloride is an orally active H1 receptor antagonist that can also activate κ-opioid and octapamine receptors. It increases ERK1/2 and CREB phosphorylation, and antagonizes full κ-opioid agonist and dynorphin A-induced MAPK phosphorylation. This compound modulates social and exploratory behavior, raises electroconvulsive thresholds, and is used in research for neurological diseases like depression and epilepsy.
- Orally active H1 receptor antagonist
- Activates κ-opioid receptor and octopamine receptor
- Increases ERK1/2 and CREB phosphorylation
- Antagonizes κ-opioid agonist and dynorphin A-induced MAPK phosphorylation
- Modulates social and exploratory behavior
- Raises electroconvulsive thresholds
- Used for research of neurological diseases, including depression and epilepsy