Learn More
Apexbio Technology LLC MIBEFRADIL-DIHYDROCHLORIDE
SDP
Supplier:  Apexbio Technology LLC A360525
Encompass_Preferred
Mibefradil dihydrochloride (CAS No 116666-63-8) is a calcium channel blocker primarily used in biomedical research for its ability to inhibit T-type calcium channels Originating from the benzimidazole class it s known for affecting pathways related to calcium influx in smooth muscle and cardiac tissues The drug exhibits in vitro activity in the nanomolar to low micromolar range making it valuable for detailed studies of calcium-dependent processes Researchers often employ it in cell models and animal studies to explore cardiovascular function neuronal signaling and potential therapeutic applications Mibefradil dihydrochloride is also utilized in drug screening to assess impacts on cellular calcium regulation Experimental concentrations typically depend on the specific research design ensuring adaptability across various experimental setups
Catalog No. 50-005-68073