Learn More
Grainger MITUTOYO IP65 Digital Outside Micrometer
Supplier:  Grainger 36J711
Encompass_Preferred
The Mitutoyo IP65 Digital Outside Micrometer is a precision measuring instrument for outside dimensions. It has a measuring range of 0-25mm/0-1in with a resolution of 0.001mm/0.00005in. The micrometer features an ABSOLUTE linear encoder that provides resistance to shock, contaminants, and power interruptions. It has a ratchet stop for consistent measuring force and is sealed to IP65 for protection against coolants, cutting fluids, and dust. The micrometer includes SPC output for statistical analysis.
Catalog No. NC2603671