Learn More
Selleck Chemical LLC Micafungin Sodium S4287-1mg-5
SDP
Supplier:  Selleck Chemical LLC S42871MG5
Encompass
Micafungin Sodium is an inhibitor of 1 3-beta-D-glucan synthesis used as an antifungal drug
Catalog No. 50-311-0448