Learn More
Medchemexpress LLC Micafungin sodium (FK 463 sodium) | 208538-73-2 | 98.9% | 1292.26 | 100 MG

Supplier: Medchemexpress LLC HY16321100MG
Micafungin sodium (FK 463 sodium) is an antifungal agent that works by inhibiting 1,3-beta-D-glucan synthesis. It is intended for research use only.
- Inhibits 1,3-beta-D-glucan synthesis
- Has a purity of 98.91%
- CAS number is 208538-73-2
- Molecular weight is 1292.26
- Formula is C56H70N9NaO23S
- Appearance is solid
- Color is white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.