Learn More
Cayman Chemical MIcafungInsodIum salt 10mg
SDP
Supplier:  Cayman Chemical 1800910 MG
Encompass_Preferred
An echinocandin antifungal that inhibits 1,3-β-D-glucan synthesis; has fungicidal activity against virtually all species of Candida, including those resistant to fluconazole, and is fungistatic against Aspergillus spp
Catalog No. 50-296-6825