Learn More
Med Vet International Michel Clip Forceps, Stainless Steel, 12cm, J0086, Each
Supplier:  Med Vet International JOR86
Encompass_Preferred
Can be used both for applying and removing Michel clips. Stainless steel. 12cm.
Catalog No. 50-005-84970