Learn More
Medchemexpress LLC Miconazole (nitrate) | 22832-87-7 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0454A10MM/1ML
Encompass_Preferred
Miconazole nitrate is an imidazole antifungal agent known for its ability to pass through the blood-brain barrier and its antibacterial effects. Developed by Janssen Pharmaceutica, it is commonly applied topically to treat fungal infections by inhibiting ergosterol synthesis in fungal cell membranes. This agent is also effective against certain species of Leishmania protozoa and is utilized in film developing processes like Ektachrome.
  • Imidazole antifungal agent
  • Passes through the blood-brain barrier
  • Exhibits antibacterial effects
  • Inhibits ergosterol synthesis in fungal cell membranes
  • Effective against certain species of Leishmania protozoa
  • Used in film developing processes
Catalog No. 50-004-08249