Learn More
Medchemexpress LLC Miconazole nitrate | 22832-87-7 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0454A500MG
Encompass_Preferred
Miconazole nitrate is an imidazole antifungal agent that can pass through the blood-brain barrier and also exhibits antibacterial effects. It works by inhibiting the synthesis of ergosterol, a crucial component of fungal cell membranes.
  • Inhibits ergosterol synthesis in fungal cell membranes
  • Exhibits antifungal and antibacterial effects
  • Can be used against certain species of Leishmania protozoa
  • Passes through the blood-brain barrier
  • Used in film developing processes
Catalog No. 50-004-09341