Learn More
Research Products International Corp Miconazole Nitrate, 25 Grams
SDP
Supplier:  Research Products International Corp M8102025.0
Encompass_Preferred
Miconazole Nitrate is an antifungal medication. It belongs to the class of azole antifungal agents and is used to combat various fungal infections. It works by inhibiting the synthesis of ergosterol, a crucial component of fungal cell membranes. Without sufficient ergosterol, the integrity of the fungal cell membrane is compromised, leading to cell death.
In research and laboratory settings, miconazole nitrate may be used as a tool to study fungal biology and the mechanisms of antifungal action. Researchers may explore its efficacy against specific fungal strains and its impact on fungal cell structures and functions.
Catalog No. 50-213-515