Learn More
Alkali Scientific Miconazole Nitrate, 5 G
SDP
Supplier:  Alkali Scientific MIC002CP
Encompass_Preferred
Miconazole Nitrate, a broad-spectrum antifungal, is available in a 5g powder form for use in laboratory and clinical applications. It effectively combats infections caused by fungi like Candida and dermatophytes by inhibiting ergosterol synthesis, essential for fungal cell membrane integrity. Miconazole is commonly used in experimental settings to test antifungal efficacy, study drug resistance, and explore mechanisms of action in fungal infections. The powder form allows researchers to prepare precise concentrations for controlled in vitro experiments, aiding in the development of new antifungal therapies and understanding resistance mechanisms in clinical isolates.
Catalog No. 50-006-59981