Learn More
Medchemexpress LLC Miconazole nitrate (Standard) | 22832-87-7 | 100.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0454AR50MG
Encompass_Preferred
Miconazole nitrate (Standard) is an analytical standard of Miconazole nitrate intended for research and analytical applications. Also known as R18134 nitrate, it is an imidazole antifungal agent that also exhibits antibacterial effects.
  • Analytical standard for research and analytical applications
  • Acts as an imidazole antifungal agent
  • Exhibits antibacterial properties
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS studies
Catalog No. 50-004-06326