Learn More
Cayman Chemical MIconazolenItrate 5g
SDP
Supplier:  Cayman Chemical 154205 G
Encompass_Preferred
A widely used antifungal imidazole that interferes with ergosterol synthesis in fungal cell membranes; induces reactive oxygen species in fungal biofilms, demonstrating a MIC value of ≥256 μM against Candida spp
Catalog No. 50-297-1988