Learn More
REPLIGEN CORPORATION Repligen MicroKros 20cm 50kD PS, High-Performance Ultrafilter, Sterile Condition
SDP
Supplier:  REPLIGEN CORPORATION C02S05005S
Encompass
The Repligen MicroKros Hollow Fiber Filter Module is a high-performance ultrafiltration device that provides precise molecular separation for scientific applications. Its 50 kD polysulfone membrane and 20 cm effective length enable efficient processing of small to medium-volume samples. Designed for use in sensitive laboratory settings, it is ready for immediate application, ensuring reliable operation for cell-free solutions while maintaining sterility for optimum performance.
Catalog No. NC2074989