Learn More
Cole-Parmer Cole-Parmer Micro-90 cleaning solution, 1 liter bottle
Supplier:  Cole-Parmer 1810005
Encompass_Preferred
Micro-90 is a mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications. Solution cleans glass, most metals, plastic, rubber, fabric, and is safe for all your ultrasonic cleaners. Radioactive contaminants will not reduce cleaning capability. Solution works at any concentration and rinses clean; most applications require only 1.25 to 2.5 fluid ounces per gallon of water. Solution is environmentally friendly—contains no phosphates, CFCs, ODCs, or borates. It safely replaces chromic acid to remove protein from glassware. Designed for use in CIP, ultrasonic and immersion applications. Can be used in validated processses. A 1% to 2% solution is effective in most applications. Typical pH: 9.5. NSF-Registered for USDA A1 use. Note: Do not use on zinc, magnesium, or aluminum. 18100-30 shipped FOB Burlington, NJ, USA.
Catalog No. NC0248628