Learn More
Braintree Scientific Micro Cannulation System .016OD x .008ID Tip w/Stylet
SDP
Supplier:  Braintree Scientific MC28
Encompass
The Micro Cannula System provides quick and convenient access to an artery or vein in small animals such as mice and rats. The tapered design of the PFA tubing provides strength while maintaining a very small distal tip of 0.016" OD x 0.008" ID. The sharpened stylette and beveled tip provides easy insertion into any vessel. Total length is approximately 10cm not including the stylette. **Sold for research purposes only and not for human use**
Catalog No. NC0427305