Learn More
World Precision Instrument Micro cleaning Brushes
SDP
Supplier:  World Precision Instrument 503231
Encompass
  • Vial of 100
  •  Non-linting, non-absorbent 2mm diameter fiber head has a patented spherical shape that holds 1/8 drop of material
  • Bendable 4" plastic handle.
Catalog No. 50-252-1281