Learn More
Roboz Surgical Micro-fiber instrument cleaning brushes, 100/pk
SDP
Supplier:  Roboz Surgical IC1002
Encompass
Micro-fiber instrument cleaning brushes. Non-absorbent, non-linting fibers, 2.0mm head size, overall length 4"
Catalog No. NC1396910