Learn More
Electron Microscopy Sciences Micro Cutting Tool Set
SDP
Supplier:  Electron Microscopy Sciences 6209302
Encompass
Includes tools #2 Micro-graver, 3 sided; #3 Micro-chisel; #4 Micro-spade; #8 Micro-knife 20 deg ; #13 Micro-graver, oval; #21 Micro-scraper, triangular; #28 Micro-scriber, carbide; MT-1 Handle; MT-case.
Catalog No. 50-996-670