Learn More
Roboz Surgical MICRO DISS SCS 4.5IN STR S/S
SDP
Supplier:  Roboz Surgical RS5912SC
Encompass
Micro Dissecting Scissors; Sure Cut Blades; Straight; Sharp Points; 24mm Blade Length; 4-1/2 in. Overall Length
Catalog No. NC9785249