Learn More
Electron Microscopy Sciences MicroPoint™ Micro Dissecting Scissors Vertical Opening, Angled 45° Blade
SDP
Supplier:  Electron Microscopy Sciences 7793003
Encompass_Preferred
20 gauge stainless steel, with a 3 mm cutting edge.
Blade: Angled 45°
Catalog No. 50-298-14