Learn More
INDUSTRIAL PROTECTION PRODUCTS INC Micro Flow Tubing for DG/DT Pump Heads, with Straps. 1x0.92 (20m)
Supplier:  INDUSTRIAL PROTECTION PRODUCTS INC 1X0.92S
Encompass_Preferred
Excellent durability and aging resistance; outstanding physical and mechanical properties; weak performance in acid and solvent resistance. Advantages: high anti-break intensity; good transparency; excellent physical and biologic inertia. Water pressure resistance: 2bar.
Catalog No. NC3789464