Learn More
Alkali Scientific ASI™ Immunoplate, PS, Clear, 96-Well, ELISA Maxibinding, Flat Bottom with Lid, 100/cs
SDP
Supplier:  Alkali Scientific MPMAXI
Encompass
Clear polystyrene. Surface chemistry for optimal binding is hydrophilic and ideal for protein research and antibody sandwich assays. Alphanumerically labeled rows and columns. Maxibinding : Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions. (Recommended for most ELISA experiments)
Catalog No. 50-238-8848