Learn More
Norgen Biotek Corp microRNA Purification Kit
Supplier:  Norgen Biotek Corp 21300
Encompass_Preferred
This kit provides a rapid method for the isolation and purification of small RNA molecules (< 200 nt) from cultured animal cells, small tissue samples, bacterial cells, plants and blood.Two columns are provided with this kit. The first column captures large RNA, while the small RNA are captured on a second column and are eluted concentrated in 25 µL of nuclease-free water.The small RNA can be used in various downstream applications relating to miRNA profiling, gene regulation and functional analysis. The eluted RNA is ready for RT-qPCR, microarrays, and NGS applications.
Catalog No. NC0698910