Learn More
Cole-Parmer Cole-Parmer Micro Stir Bar, 5 mm L x 2 mm Dia; 1/Pk
Supplier:  Cole-Parmer 0854501
Encompass_Preferred
Strong alnico V magnets with one-piece coating of PTFE.
Catalog No. NC9028779