Learn More
Electron Microscopy Sciences Micro-Tip Cryo-Markers™, Black
SDP
Supplier:  Electron Microscopy Sciences 6205502BK
Encompass_Preferred
High quality cryogenic permanent ink markers. High quality permanent ink micro-tip marker with ultra-fine line 0.25mm. The hard plastic tips are protected by metal sleeves and will not split, so lines are clean and precise every time. All colors are equally permanent and water-resistant. Excellent performance in areas where detailed writing is required, such as marking on small micro-centrifuge and PCR tubes, labels, beakers, glass plates, X-ray and photographic films.
Color: Black
Catalog No. NC1511072