Learn More
Quidel Corporation MICROVUE CIC-C1Q ELISA KIT
SDP
Supplier:  Quidel Corporation A001
Encompass
The MicroVue CIC-C1q EIA is designed for detection of C1q binding circulating immune complexes in human serum and plasma.
Catalog No. NC1622987