Learn More
Electron Microscopy Sciences Microbeaker Holder
SDP
Supplier:  Electron Microscopy Sciences 72645
Encompass_Preferred
Brushed aluminum rack holds six 12 mm O.D. microbeakers in unique, stepped "see-through" holes for illuminated and microscopic observation of samples. Holds six 12mm OD microbeakers.
Catalog No. 50-949-300