Learn More
Enzo Life Sciences Microcystin-HilR (25µg)
SDP
Supplier:  Enzo Life Sciences ALX350177C025
Encompass_Preferred
Isolated from Microcystis aeruginosa. Microcystin containing a homoisoleucine (Hil) in position 2 and arginine (R) in position 4. Hepatotoxic. Alternative name: [Hil2]microcystin-IR. Purity: ≥95% (HPLC). Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8776